6-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine 95%

Supplier: Ambeed
Warning

A317336-100MG A317336-50MG
77015-226EA 124.13 USD
77015-226 77015-228
6-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine 95%
6-Bromo-3-iodo-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrIN₃
MW: 323.92 g/mol
MDL Number: MFCD22380440
CAS Number: 1357946-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR