Methyl 2-methylpyrazole-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-1H-pyrazole-5-carboxylate

A317163-10G A317163-1G A317163-25G A317163-5G
76989-060EA 95.49 USD
76989-060 76989-062 76989-064 76989-066
Methyl 2-methylpyrazole-3-carboxylate 98%
Methyl 2-methylpyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD04967357
CAS Number: 17827-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR