4-(1H-Pyrazol-1-yl)benzoic acid 96%

Supplier: Ambeed
Warning

A316887-250MG A316887-5G
77015-216EA 8.49 USD
77015-216 77015-218
4-(1H-Pyrazol-1-yl)benzoic acid 96%
4-(1H-Pyrazol-1-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.18 g/mol
MDL Number: MFCD02682056
CAS Number: 16209-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR