Acetato(2'-di-t-butylphosphino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2'-Di-tert-butylphosphine)biphenylpalladium(II) acetate

A316878-100MG A316878-1G A316878-250MG
77015-210EA 39.26 USD
77015-210 77015-212 77015-214
Acetato(2'-di-t-butylphosphino-1,1'-biphenyl-2-yl)palladium(II) 98%
Acetato(2'-di-t-butylphosphino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₂₂H₂₉O₂PPd
MW: 462.8600 g/mol
MDL Number: MFCD04117716
CAS Number: 577971-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR