3-Isobutyl-1-methyl-1H-pyrazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A316857-1G A316857-250MG A316857-25G A316857-5G
76989-056EA 20 USD
76989-056 76989-058 77890-624 77890-625
3-Isobutyl-1-methyl-1H-pyrazole-5-carboxylic acid 95%
3-Isobutyl-1-methyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂
MDL Number: MFCD05670720
CAS Number: 769132-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR