Methyl 3-bromo-4-isopropoxybenzoate 95%

Supplier: Ambeed
Warning

A316611-100MG A316611-250MG A316611-5G
76989-050EA 8 USD
76989-050 76989-052 76989-054
Methyl 3-bromo-4-isopropoxybenzoate 95%
Methyl 3-bromo-4-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₃
MW: 273.12 g/mol
MDL Number: MFCD09271835
CAS Number: 213598-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR