2-Chloro-5-nitrobenzoic acid 99%

Supplier: Ambeed
Warning

A316605-100G A316605-25G
77002-786EA 18.04 USD
77002-786 77002-788
2-Chloro-5-nitrobenzoic acid 99%
2-Chloro-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
MDL Number: MFCD00007294
CAS Number: 2516-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR