Benzo[b]naphtho[1,2-d]furan 97%

Supplier: Ambeed
Danger

A316594-100MG A316594-1G A316594-250MG A316594-5G
77032-700EA 18 USD
77032-700 77032-702 77032-704 77032-706
Benzo[b]naphtho[1,2-d]furan 97%
Benzo[b]naphtho[1,2-d]furan

Precaución: for Research Use Only

Formula: C₁₆H₁₀O
MDL Number: MFCD00046916
CAS Number: 205-39-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR