Methyl 4-chloro-3-methoxybenzoate 98%

Supplier: Ambeed
Warning

A316423-25G A316423-5G
77015-206EA 84 USD
77015-206 77015-208
Methyl 4-chloro-3-methoxybenzoate 98%
Methyl 4-chloro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD02683861
CAS Number: 116022-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR