5-Iodo-6-(trifluoromethyl)pyridin-2-ol 95%

Supplier: Ambeed
Warning

A316377-100MG A316377-1G
77015-202EA 9 USD
77015-202 77015-204
5-Iodo-6-(trifluoromethyl)pyridin-2-ol 95%
5-Iodo-6-(trifluoromethyl)pyridin-2-ol

Precaución: for Research Use Only

Formula: C₆H₃F₃INO
MW: 288.994 g/mol
MDL Number: MFCD16611365
CAS Number: 1227515-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR