Methyl 5-bromo-2-hydroxy-4-methoxybenzoate 95%

Supplier: Ambeed
Warning

A316365-1G A316365-250MG
76989-038EA 91.24 USD
76989-038 76989-040
Methyl 5-bromo-2-hydroxy-4-methoxybenzoate 95%
Methyl 5-bromo-2-hydroxy-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₄
MW: 261.07 g/mol
MDL Number: MFCD03426751
CAS Number: 39503-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR