(6-Chloro-pyridazin-3-yl)dimethylamine 97%

Supplier: Ambeed
Warning

A316272-1G A316272-250MG
77032-696EA 43 USD
77032-696 77032-698
(6-Chloro-pyridazin-3-yl)dimethylamine 97%
(6-Chloro-pyridazin-3-yl)dimethylamine

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
MDL Number: MFCD02732844
CAS Number: 7145-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR