(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-phenylbutanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-β-HoPhe-OH

A316154-1G A316154-250MG
77002-776EA 102.91 USD
77002-776 77002-778
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-phenylbutanoic acid 97%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₄
MW: 401.45 g/mol
MDL Number: MFCD04117843
CAS Number: 209252-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR