3-Bromo-5-nitropyridin-4-ol 95%

Supplier: Ambeed
Warning

A315443-1G A315443-250MG A315443-5G A315443-25G
77002-764EA 11 USD
77002-764 77002-766 77002-768 77593-878
3-Bromo-5-nitropyridin-4-ol 95%
3-Bromo-5-nitropyridin-4-ol

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₃
MDL Number: MFCD08692339
CAS Number: 31872-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR