2-Bromo-N-(2-methoxyphenyl)acetamide 95%

Supplier: Ambeed
Warning

A315121-100MG A315121-250MG A315121-5G A315121-1G
77032-686EA 19 USD
77032-686 77032-688 77032-690 77872-441
2-Bromo-N-(2-methoxyphenyl)acetamide 95%
2-Bromo-N-(2-methoxyphenyl)acetamide
Formula: C₉H₁₀BrNO₂
MDL Number: MFCD02974384
CAS Number: 32428-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR