3,5-Dibromopyridin-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromo-4-hydroxypyridine

A314956-1G A314956-25G A314956-5G
77002-754EA 19.1 USD
77002-754 77002-756 77002-758
3,5-Dibromopyridin-4-ol 98%
3,5-Dibromopyridin-4-ol

Precaución: for Research Use Only

Formula: C5H3Br2NO
MW: 252.8914 g/mol
MDL Number: MFCD00082969
CAS Number: 25813-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR