Ethyl (2-fluorophenyl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-fluorophenylacetate

A314871-100G A314871-1G A314871-5G
77032-680EA 101 USD
77032-680 77032-682 77032-684
Ethyl (2-fluorophenyl)acetate 98%
Ethyl (2-fluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
MDL Number: MFCD04039343
CAS Number: 584-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR