2-Chloro-3-cyclopropylpyridine 95%

Supplier: Ambeed
Warning

A314752-100MG A314752-1G A314752-250MG A314752-5G
76989-016EA 50.93 USD
76989-016 76989-018 76989-020 76989-022
2-Chloro-3-cyclopropylpyridine 95%
2-Chloro-3-cyclopropylpyridine

Precaución: for Research Use Only

Formula: C₈H₈ClN
MW: 153.60 g/mol
MDL Number: MFCD18254227
CAS Number: 865664-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR