1-Boc-4-(Aminomethyl)piperidine hydrochloride 97%

Supplier: Ambeed
Warning

A314359-1G A314359-250MG A314359-25G A314359-5G
77032-662EA 23 USD
77032-662 77032-664 77032-666 77032-668
1-Boc-4-(Aminomethyl)piperidine hydrochloride 97%
1-Boc-4-(Aminomethyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₃ClN₂O₂
MDL Number: MFCD03093510
CAS Number: 359629-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR