2,3-Dichloro-4-methyl-5-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Danger

A314337-1G A314337-250MG
77015-194EA 303 USD
77015-194 77015-196
2,3-Dichloro-4-methyl-5-(trifluoromethyl)pyridine 95%
2,3-Dichloro-4-methyl-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₄Cl₂F₃N
MDL Number: MFCD27939589
CAS Number: 431942-49-3
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR