Methyl-2-fluoro-4-hydroxybenzoate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 2-fluoro-4-hydroxybenzoate
A314124-1G
A314124-250MG
A314124-5G
A314124-100G
A314124-10G
A314124-25G
77002-740EA
6.37
USD
77002-740
77002-742
77002-744
77002-746
77672-608
77881-877
77881-878
Methyl-2-fluoro-4-hydroxybenzoate 98%
Methyl-2-fluoro-4-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₈H₇FO₃ |
MDL Number:
MFCD09038282 CAS Number: 197507-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...