5-Bromo-2-methox-4-methylypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methoxy-4-methylpyridine

A314109-100G A314109-1G A314109-25G A314109-5G
77015-186EA 384.05 USD
77015-186 77015-188 77015-190 77015-192
5-Bromo-2-methox-4-methylypyridine 98%
5-Bromo-2-methox-4-methylypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.0485 g/mol
MDL Number: MFCD04039980
CAS Number: 164513-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR