(S)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole ≥98%, ee 99%

Supplier: Ambeed
Warning

A313682-1G A313682-25G A313682-5G A313682-250MG
76989-010EA 46 USD
76989-010 76989-012 76989-014 77593-872
(S)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole ≥98%, ee 99%
(S)-5,5'-Bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole

Precaución: for Research Use Only

Formula: C₃₈H₂₈O₄P₂
MDL Number: MFCD09753005
CAS Number: 210169-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR