(Tetrahydrofuran-3-yl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Aminomethyltetrahydrofuran hydrochloride, Oxolan-3-ylmethanamine hydrochloride, 3-(Aminomethyl)oxolane hydrochloride

A313498-10G A313498-1G A313498-5G
77015-180EA 79.57 USD
77015-180 77015-182 77683-930
(Tetrahydrofuran-3-yl)methanamine hydrochloride 97%
(Tetrahydrofuran-3-yl)methanamine hydrochloride

Precaución: For research use only.

Formula: C₅H₁₂ClNO
MW: 137.61 g/mol
MDL Number: MFCD08448154
CAS Number: 184950-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR