1-(((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)(piperidin-1-yl)methylene)piperidin-1-ium hexafluorophosphate(V) 98%
Supplier: Ambeed
|
Danger
|
A313335-1G
A313335-25G
A313335-5G
77002-726EA
19.1
USD
77002-726
77002-728
77002-730
1-(((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)(piperidin-1-yl)methylene)piperidin-1-ium hexafluorophosphate(V) 98%
1-(((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)(piperidin-1-yl)methylene)piperidin-1-ium hexafluorophosphate(V)
Precaución: for Research Use Only
Formula:
C₁₇H₂₄F₆N₅OP MW: 459.37 g/mol |
MDL Number:
MFCD00191769 CAS Number: 206752-41-2 UN: 1325 ADR: 4.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...