2-Hydroxypyrimidine-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A313108-100MG A313108-1G
77032-646EA 102.91 USD
77032-646 77032-648
2-Hydroxypyrimidine-5-carbaldehyde 95%
2-Hydroxypyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₄N₂O₂
MW: 124.10 g/mol
MDL Number: MFCD10696897
CAS Number: 166757-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR