5-Bromo-4-(trifluoromethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A313043-100MG A313043-5G A313043-250MG
77032-642EA 29.71 USD
77032-642 77032-644 77601-410
5-Bromo-4-(trifluoromethyl)pyrimidine 97%
5-Bromo-4-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₂BrF₃N₂
MW: 226.9820 g/mol
MDL Number: MFCD11046325
CAS Number: 785777-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR