(±)-2-Methyltetrahydrofuran-3-one 98%

Supplier: Ambeed
Danger

A312941-10G A312941-25G
77002-720EA 11 USD
77002-720 77672-598
(±)-2-Methyltetrahydrofuran-3-one 98%
(±)-2-Methyltetrahydrofuran-3-one
Formula: C₅H₈O₂
MDL Number: MFCD00010423
CAS Number: 3188-00-9
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR