7-Deaza-7-iodo-2'-deoxyguanosine 95%

Supplier: Ambeed
Warning

A312793-250MG A312793-1G A312793-5G A312793-100MG
76989-004EA 94 USD
76989-004 77571-874 77571-876 77612-798
7-Deaza-7-iodo-2'-deoxyguanosine 95%
7-Deaza-7-iodo-2'-deoxyguanosine

Precaución: for Research Use Only

Formula: C₁₁H₁₃IN₄O₄
MDL Number: MFCD07778651
CAS Number: 172163-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR