1,4-Di(1-pyrenyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 1,1'-(1,4-Phenylene)bispyrene, 1,4-Di(pyren-1-yl)benzene

A312478-1G A312478-250MG
77015-168EA 71.09 USD
77015-168 77015-170
1,4-Di(1-pyrenyl)benzene 98%
1,4-Di(1-pyrenyl)benzene

Precaución: for Research Use Only

Formula: C₃₈H₂₂
MW: 478.59 g/mol
MDL Number: MFCD12022457
CAS Number: 475460-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR