1-(2,2,2-Trifluoroethyl)piperidin-4-one 97%

Supplier: Ambeed
Warning

A312222-100MG A312222-5G A312222-1G
77015-166EA 33.95 USD
77015-166 77612-796 77683-921
1-(2,2,2-Trifluoroethyl)piperidin-4-one 97%
1-(2,2,2-Trifluoroethyl)piperidin-4-one

Precaución: For research use only.

Formula: C₇H₁₀F₃NO
MW: 181.156 g/mol
MDL Number: MFCD09936907
CAS Number: 81363-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR