3-Chloro-2-fluoro-5-(trifluoromethyl)benzonitrile 97%

Supplier: Ambeed
Warning

A312194-100MG A312194-250MG A312194-1G A312194-5G
76988-982EA 49 USD
76988-982 76988-984 77890-608 77890-609
3-Chloro-2-fluoro-5-(trifluoromethyl)benzonitrile 97%
3-Chloro-2-fluoro-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₂ClF₄N
MDL Number: MFCD09999623
CAS Number: 129931-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR