3-Chloro-4-hydroxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-4-hydroxyphenyl)boronic acid

A312011-100MG A312011-10G A312011-1G A312011-250MG A312011-5G
77032-624EA 7.43 USD
77032-624 77032-626 77032-628 77032-630 77032-632
3-Chloro-4-hydroxyphenylboronic acid 98%
3-Chloro-4-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BClO₃
MW: 172.3700 g/mol
MDL Number: MFCD09258739
CAS Number: 182344-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR