tert-Butyl N-(1,3-oxazol-4-yl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl oxazol-4-ylcarbamate

A311930-250MG A311930-1G A311930-5G
76988-980EA 19 USD
76988-980 77890-606 77890-607
tert-Butyl N-(1,3-oxazol-4-yl)carbamate 95%
tert-Butyl N-(1,3-oxazol-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₃
MDL Number: MFCD18533643
CAS Number: 1314931-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR