Cyclohexane-1,2-diamine, 1-BOC protected 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl (2-aminocyclohex-1-yl)carbamate, tert-Butyl (2-aminocyclohexyl)carbamate, 1-Amino-2-[(tert-butoxycarbonyl)amino]cyclohexane
A311906-250MG
A311906-1G
77002-696EA
14
USD
77002-696
77881-870
Cyclohexane-1,2-diamine, 1-BOC protected 98%
Cyclohexane-1,2-diamine, 1-BOC protected
Precaución: for Research Use Only
Formula:
C₁₁H₂₂N₂O₂ |
MDL Number:
MFCD04038473 CAS Number: 317595-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...