2-Chloro-4-ethoxy-6-methylpyrimidine 95%

Supplier: Ambeed
Warning

A311854-1G A311854-250MG A311854-5G
77002-690EA 79 USD
77002-690 77002-692 77002-694
2-Chloro-4-ethoxy-6-methylpyrimidine 95%
2-Chloro-4-ethoxy-6-methylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
MDL Number: MFCD00234338
CAS Number: 37482-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR