Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate 97%

Supplier: Ambeed
Warning

A311178-10G A311178-1G A311178-5G A311178-250MG
77032-606EA 91 USD
77032-606 77032-608 77032-610 77872-431
Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate 97%
Ethyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₂O₃S
MDL Number: MFCD09833701
CAS Number: 160844-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR