2-Amino-4-oxo-1,4-dihydropyrimidine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A310912-100MG A310912-1G A310912-250MG
77032-600EA 151 USD
77032-600 77032-602 77032-604
2-Amino-4-oxo-1,4-dihydropyrimidine-5-carbonitrile 95%
2-Amino-4-oxo-1,4-dihydropyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄N₄O
MW: 136.11 g/mol
MDL Number: MFCD16621665
CAS Number: 27058-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR