L-Leucine ethyl ester hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Leu-OEt.HCl
A310551-100G
A310551-25G
A310551-500G
77002-672EA
41
USD
77002-672
77002-674
77002-676
77002-678
L-Leucine ethyl ester hydrochloride 97%
L-Leucine ethyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₇NO₂ MW: 195.69 g/mol |
MDL Number:
MFCD00034879 CAS Number: 2743-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...