2-Methyl-2,7-diazaspiro[3.5]nonane dihydrochloride 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A310240-100MG A310240-1G A310240-250MG
77015-152EA 82.76 USD
77015-152 77015-154 77683-915
2-Methyl-2,7-diazaspiro[3.5]nonane dihydrochloride 95%, Ambeed.Inc
2-Methyl-2,7-diazaspiro[3.5]nonane dihydrochloride

Precaución: For research use only.

Formula: C₈H₁₈Cl₂N₂
MW: 213.14 g/mol
MDL Number: MFCD26406720
CAS Number: 1610028-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR