(4-Chlorophenyl)(1H-imidazol-2-yl)methanone 95%

Supplier: Ambeed
Warning

A310168-1G A310168-250MG
77015-148EA 101.85 USD
77015-148 77015-150
(4-Chlorophenyl)(1H-imidazol-2-yl)methanone 95%
(4-Chlorophenyl)(1H-imidazol-2-yl)methanone

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂O
MW: 206.62 g/mol
MDL Number: MFCD04110656
CAS Number: 62457-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR