7-Fluoro-6-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A310098-100MG A310098-1G A310098-250MG
76988-948EA 47.75 USD
76988-948 76988-950 76988-952
7-Fluoro-6-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
7-Fluoro-6-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₀ClFN₂O₂
MW: 232.63 g/mol
MDL Number: MFCD18157610
CAS Number: 912846-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR