tert-Butyl-3-phenylpiperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Phenylpiperazine, N1-BOC protected, 1-Boc-3-Phenylpiperazine

A310063-100MG A310063-1G A310063-250MG A310063-5G
77032-582EA 24.41 USD
77032-582 77032-584 77032-586 77032-588
tert-Butyl-3-phenylpiperazine-1-carboxylate 98%
tert-Butyl-3-phenylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₂
MW: 262.35 g/mol
MDL Number: MFCD03840082
CAS Number: 502649-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR