4-(Bromomethyl)-N,N-diphenylaniline 95%

Supplier: Ambeed
Danger

A309994-100MG A309994-250MG
76988-944EA 149 USD
76988-944 76988-946
4-(Bromomethyl)-N,N-diphenylaniline 95%
4-(Bromomethyl)-N,N-diphenylaniline

Precaución: for Research Use Only

Formula: C₁₉H₁₆BrN
MDL Number: MFCD20485908
CAS Number: 183994-94-7
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR