6-Bromoisoquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A309784-100MG A309784-250MG A309784-5G A309784-1G
76988-936EA 62 USD
76988-936 76988-938 76988-940 77593-860
6-Bromoisoquinoline-3-carboxylic acid 97%
6-Bromoisoquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MDL Number: MFCD22689652
CAS Number: 1416713-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR