4-Nitropyridine 95%

Supplier: Ambeed
Warning

A309598-1G A309598-250MG A309598-5G
77015-140EA 56.23 USD
77015-140 77015-142 77015-144
4-Nitropyridine 95%
4-Nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄N₂O₂
MW: 124.097 g/mol
MDL Number: MFCD00054554
CAS Number: 1122-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR