2-Chloro-4-methoxy-3-nitropyridine 97%
Supplier: Ambeed
|
Warning
|
A309444-1G
A309444-250MG
A309444-5G
77032-578EA
12
USD
77032-578
77032-580
77668-370
2-Chloro-4-methoxy-3-nitropyridine 97%
2-Chloro-4-methoxy-3-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O₃ |
MDL Number:
MFCD00955756 CAS Number: 6980-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...