5-Hydroxypiperidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A309315-100MG A309315-250MG A309315-1G A309315-5G
76988-924EA 54 USD
76988-924 76988-926 77593-852 77593-854
5-Hydroxypiperidine-2-carboxylic acid 95%
5-Hydroxypiperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃
MW: 145.15 g/mol
MDL Number: MFCD08694639
CAS Number: 13096-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR