2-Bromo-3-(difluoromethoxy)pyridine 95%

Supplier: Ambeed
Warning

A309203-100MG A309203-250MG A309203-5G A309203-1G
77015-134EA 64.72 USD
77015-134 77015-136 77015-138 77568-622
2-Bromo-3-(difluoromethoxy)pyridine 95%
2-Bromo-3-(difluoromethoxy)pyridine

Precaución: For research use only.

Formula: C₆H₄BrF₂NO
MW: 224.003 g/mol
MDL Number: MFCD13185840
CAS Number: 947249-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR