(S)-[(1-Phenylethyl)amino]acetic acid 97%

Supplier: Ambeed
Warning

A309101-10G A309101-1G A309101-5G
77002-666EA 52 USD
77002-666 77002-668 77002-670
(S)-[(1-Phenylethyl)amino]acetic acid 97%
(S)-[(1-Phenylethyl)amino]acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD11656778
CAS Number: 78397-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR